Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromotoluene ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

220115-100G 220115-25G 220115-500G
BT220115-100GEA 49 USD InStock
BT220115-100G BT220115-25G BT220115-500G
4-Bromotoluene ≥98%
4-Bromotoluene
CAS: 106-38-7; EC No: 203-391-8; MDL No: MFCD00000109; RTECS: XS7965600 Solid; Molecular Formula: C7H7Br ; MW: 171.03 Melting Point: 26-29°; Boiling Point: 184°; Flash point: 85°C (185°F) Density (g/mL): 1.39

  • RTECS: XS7965600
  • Solid


Formula: C₇H₇Br
Boiling Pt: 184 °C
Melting Pt: 26…29 °C
Density: 1.39 g/mL
Flash Pt: 85 °C (185 °F)
MDL Number: MFCD00000109
CAS Number: 106-38-7
EINECS: 203-391-8
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty