Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromoacenaphthene ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

220150-25G 220150-5G
BT220150-25GEA 228 USD InStock
BT220150-25G BT220150-5G
5-Bromoacenaphthene ≥97%
5-Bromoacenaphthene
CAS: 2051-98-1; EC No: 218-138-7; MDL No: MFCD00003809 Powder; Molecular Formula: C12H9Br; MW: 233.10 Melting Point: 50-56°; Boiling Point: 335°

  • Powder


Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 335
Melting Pt: 54
MDL Number: MFCD00003809
CAS Number: 2051-98-1
EINECS: 218-138-7
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty