Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorothiazole ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

220460-1G 220460-5G
BT220460-1GEA 161 USD InStock
BT220460-1G BT220460-5G
2-Chlorothiazole ≥97%
2-Chlorothiazole
CAS: 3034-52-4; EC No: 221-228-9; MDL No: MFCD00210701 Liquid; Molecular Formula: C3H2ClNS; MW: 119.57 Density (g/mL): 1.393; Refractive Index: 1.551

  • Refractive index: 1.551
  • Liquid


Formula: C₃H₂ClNS
MW: 119.57
Density: 1.393 g/mL
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty