Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2',3'-Difluoroacetophenone ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

220610-1G 220610-25G 220610-5G
BT220610-1GEA 49 USD InStock
BT220610-1G BT220610-25G BT220610-5G
2',3'-Difluoroacetophenone ≥97%
2',3'-Difluoroacetophenone
CAS: 18355-80-1; MDL No: MFCD00061274 Liquid; Molecular Formula: C8H6F2O; MW: 156.13

  • Liquid


Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD00061274
CAS Number: 18355-80-1
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty