Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroisonicotinic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

220980-1G 220980-5G
BT220980-1GEA 30 USD InStock
BT220980-1G BT220980-5G
2-Chloroisonicotinic acid ≥98%
2-Chloroisonicotinic acid
CAS: 6313-54-8; MDL No: MFCD00191402 Solid; Molecular Formula: C6H3ClNO2; MW: 157.55 Melting Point: 246° (decomposes)

  • Solid


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 246 °C (dec.)
MDL Number: MFCD00191402
CAS Number: 6313-54-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty