Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Dap(Fmoc)-OH ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

221595-1G 221595-5G
BT221595-1GEA 94 USD InStock
BT221595-1G BT221595-5G
Boc-Dap(Fmoc)-OH ≥98%
Boc-Dap(Fmoc)-OH
CAS: 122235-70-5; MDL No: MFCD00235897 Powder; Molecular Formula: C23H26N2O6; MW: 426.46

  • Powder


Formula: C₂₃H₂₆N₂O₆
MW: 426.469 g/mol
MDL Number: MFCD00235897
CAS Number: 122235-70-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty