Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium diphenylamine-4-sulphonate ACS

Supplier: BEANTOWN CHEMICAL - BTC

221780-25G 221780-5G
BT221780-25GEA 153 USD InStock
BT221780-25G BT221780-5G
Sodium diphenylamine-4-sulphonate ACS
Sodium diphenylamine-4-sulphonate
CAS: 6152-67-6; EC No: 228-165-6; MDL No: MFCD00036468 Powder; Molecular Formula: C12H10NNaO3S; MW: 271.27 Moisture sensitive

  • Moisture sensitive
  • Powder


Formula: C₁₂H₁₀NNaO₃S
MW: 271.27 g/mol
MDL Number: MFCD00036468
CAS Number: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty