Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Abietic acid ≥75%, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

222090-100G 222090-25G
BT222090-100GEA 131 USD InStock
BT222090-100G BT222090-25G
Abietic acid ≥75%, Technical Grade
Abietic acid
CAS: 514-10-3; EC No: 208-178-3; MDL No: MFCD03423567; RTECS: TP8580000 Crystalline; Molecular Formula: C20H30O2; MW: 302.45 Melting Point: 139-142° Air Sensitive

  • RTECS: TP8580000
  • Air Sensitive
  • Crystalline


Formula: C₂₀H₃₀O₂
Melting Pt: 139…142 °C
MDL Number: MFCD03423567
CAS Number: 514-10-3
EINECS: 208-178-3
Order Now
Specification Test Results
Assay 75%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty