Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiophene-3-carbaldehyde ≥96%

Supplier: BEANTOWN CHEMICAL - BTC

222115-10G 222115-250G 222115-50G
BT222115-10GEA 76 USD InStock
BT222115-10G BT222115-250G BT222115-50G
Thiophene-3-carbaldehyde ≥96%
Thiophene-3-carbaldehyde
CAS: 498-62-4; EC No: 207-865-5; MDL No: MFCD00005466 Liquid; Molecular Formula: C5H4OS; MW: 112.15 Boiling Point: 194-196°; Flash point: 64°C (147°F) Density (g/mL): 1.28; Refractive Index: 1.583 Air Sensitive

  • Refractive index: 1.583
  • Air Sensitive
  • Liquid


Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 191
Melting Pt: -30
Density: 1.28 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5
Order Now
Specification Test Results
Assay 96%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty