Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium oxide, γ-phase ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

222795-100G 222795-25G 222795-500G
BT222795-100GEA 169 USD InStock
BT222795-100G BT222795-25G BT222795-500G
Aluminium oxide, γ-phase ≥99%
Aluminium oxide, γ-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 40-80nm APS Powder, S. A. 180 m2/g; Molecular Formula: Al2O3 ; MW: 101.96 Melting Point: 2040°

  • 40-80nm APS Powder, S. A. 180 m²/g


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty