Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glutaric anhydride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

222880-100G 222880-250G
BT222880-100GEA 65 USD InStock
BT222880-100G BT222880-250G
Glutaric anhydride ≥98%
Glutaric anhydride
CAS: 108-55-4; EC No: 203-593-6; MDL No: MFCD00006679; RTECS: MA3850000 Crystalline/Solid; Molecular Formula: C5H6O3; MW: 114.10 Melting Point: 50-55°; Boiling Point: 150°/10 mmHg; Flash point: 173°C (343°F) Moisture Sensitive

  • RTECS: MA3850000
  • Moisture Sensitive
  • Crystalline/Solid


Formula: C₅H₆O₃
Boiling Pt: 150 °C/10 mmHg
Melting Pt: 50…55 °C
Flash Pt: 173 °C (343 °F)
MDL Number: MFCD00006679
CAS Number: 108-55-4
EINECS: 203-593-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty