223555-250G
223555-50G
BT223555-250GEA
195
USD
InStock
BT223555-250G
BT223555-50G
4-Chloro-1-butanol ≥85%, Technical Grade
4-Chloro-1-butanol
CAS: 928-51-8; EC No: 213-175-5; MDL No: MFCD00002967; RTECS: EL1325000
Liquid; Linear Formula: Cl(CH2)4OH; MW: 108.57
Boiling Point: 84-85°/16 mmHg; Flash point: 36°C (97°F)
Density (g/mL): 1.088; Refractive Index: 1.453
- RTECS: EL1325000
- Refractive index: 1.453
- Liquid