Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

223950-1G 223950-250MG 223950-5G
BT223950-1GEA 200 USD InStock
BT223950-1G BT223950-250MG BT223950-5G
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid ≥98%
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid
CAS: 85977-52-2; MDL No: MFCD07368365 Crystalline/Powder; Molecular Formula: C11H12O2 ; MW: 176.21

  • Crystalline/Powder


Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD07368365
CAS Number: 85977-52-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty