Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxybenzaldehyde dimethyl acetal ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

223990-100G 223990-25G 223990-500G
BT223990-100GEA 123 USD InStock
BT223990-100G BT223990-25G BT223990-500G
4-Methoxybenzaldehyde dimethyl acetal ≥98%
4-Methoxybenzaldehyde dimethyl acetal
CAS: 2186-92-7; EC No: 218-577-4; MDL No: MFCD00036507 Liquid; Molecular Formula: C10H14O3; MW: 182.22 Boiling Point: 85-87°/0.1 mmHg; Flash point: 114°C (237°F) Density (g/mL): 1.07; Refractive Index: 1.505

  • Refractive index: 1.505
  • Liquid


Formula: C₁₀H₁₄O₃
Boiling Pt: 85…87 °C/0.1 mmHg
Density: 1.07 g/mL
Flash Pt: 114 °C (237 °F)
MDL Number: MFCD00036507
CAS Number: 2186-92-7
EINECS: 218-577-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty