Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridoxine hydrochloride ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

224220-100G 224220-25G 224220-500G
BT224220-100GEA 100 USD InStock
BT224220-100G BT224220-25G BT224220-500G
Pyridoxine hydrochloride ≥99%
Pyridoxine hydrochloride
CAS: 58-56-0; EC No: 200-386-2; MDL No: MFCD00012807; RTECS: UV1350000 Crystalline/Powder; Molecular Formula: C8H11NO3·HCl; MW: 205.64 Melting Point: 214-215° Light Sensitive

  • RTECS: UV1350000
  • Light Sensitive
  • Crystalline/Powder


Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Melting Pt: 214…215 °C
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty