Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Methyl-2-propanesulfinamide ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

224465-1G 224465-25G 224465-5G
BT224465-1GEA 72 USD InStock
BT224465-1G BT224465-25G BT224465-5G
(R)-(+)-2-Methyl-2-propanesulfinamide ≥98%
(R)-(+)-2-Methyl-2-propanesulfinamide
CAS: 196929-78-9; MDL No: MFCD05861479 Solid; Molecular Formula: C4H11NOS ; MW: 121.20 Melting Point: 103-107° Optical Rotation: [α]20/D +44°, c = 1.0242 in chloroform

  • Optical rotation: [α]20/D +44°, c = 1.0242 in chloroform
  • Solid


Formula: C₄H₁₁NOS
MW: 121.20 g/mol
Melting Pt: 103…107 °C
MDL Number: MFCD05861479
CAS Number: 196929-78-9
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty