Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chlorophenyl)acetic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

224605-10G 224605-250G 224605-50G
BT224605-10GEA 40 USD InStock
BT224605-10G BT224605-250G BT224605-50G
(2-Chlorophenyl)acetic acid ≥98%
(2-Chlorophenyl)acetic acid
CAS: 2444-36-2; EC No: 219-482-0; MDL No: MFCD00004317 Powder; Molecular Formula: C8H7O2Cl; MW: 170.59 Melting Point: 92-95°

  • Powder


Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Melting Pt: 92…95 °C
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty