224820-100G
224820-500G
BT224820-100GEA
35
USD
InStock
BT224820-100G
BT224820-500G
Hydantoin ≥99%
Hydantoin
CAS: 461-72-3; EC No: 207-313-3; MDL No: MFCD00005259; RTECS: MT8210000
Solid; Molecular Formula: C3H4N2O2; MW: 100.08
Melting Point: 221-223°