Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium chloride xhydrate

Supplier: BEANTOWN CHEMICAL - BTC

225120-100G 225120-25G 225120-5G
BT225120-100GEA 193 USD InStock
BT225120-100G BT225120-25G BT225120-5G
Tetrabutylammonium chloride xhydrate
Tetrabutylammonium chloride xhydrate
CAS: 37451-68-6; EC No: 214-195-7; MDL No: MFCD00011635 Crystalline; Linear Formula: [CH3(CH2)3]4N(Cl)·xH2O; Molecular Formula: C16H36ClN·xH2O; MW: 277.92 (anhydrous) Melting Point: 41-44° Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: C₁₆H₃₆ClN·xH₂O
MW: 277.92 g/mol (anhy)
Melting Pt: 41…44 °C
MDL Number: MFCD00011635
CAS Number: 37451-68-6
EINECS: 214-195-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty