Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl-2-urea ≥96%

Supplier: BEANTOWN CHEMICAL - BTC

225190-100G 225190-500G
BT225190-100GEA 217 USD InStock
BT225190-100G BT225190-500G
1-Butyl-2-urea ≥96%
1-Butyl-2-urea
CAS: 592-31-4; EC No: 209-748-4; MDL No: MFCD00007956; RTECS: YS3675000 Crystalline; Linear Formula: CH3(CH2)3NHCONH2; Molecular Formula: C5H12N2O; MW: 116.16 Melting Point: 95-98°

  • RTECS: YS3675000
  • Crystalline


Formula: C₅H₁₂N₂O
Melting Pt: 95…98 °C
MDL Number: MFCD00007956
CAS Number: 592-31-4
EINECS: 209-748-4
Order Now
Specification Test Results
Assay 96%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty