Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Octadecane ≥90%, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

225270-1KG 225270-5KG
BT225270-1KGEA 44 USD InStock
BT225270-1KG BT225270-5KG
n-Octadecane ≥90%, Technical Grade
n-Octadecane
CAS: 593-45-3; EC No: 209-790-3; MDL No: MFCD00009007 Wet Crystalline; Linear Formula: CH3(CH2)16CH3; Molecular Formula: C18H38; MW: 254.50 Melting Point: 26-29°; Boiling Point: 317°; Flash point: 166°C (331°F) Density (g/mL): 0.777; Refractive Index: 1.439

  • Refractive index: 1.439
  • Crystalline


Formula: H₃C(CH₂)₁₆CH₃
Boiling Pt: 317 °C
Melting Pt: 26…29 °C
Density: 0.777 g/mL
Flash Pt: 166 °C (331 °F)
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3
Order Now
Specification Test Results
Assay 90%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty