Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Molybdenum ≥99.95% (trace metals basis), foil, Thickness 0.127 mm

Supplier: BEANTOWN CHEMICAL - BTC

225565-100X150 225565-150X300 225565-300X600
BT225565-100X1EA 68 USD InStock
BT225565-100X1 BT225565-150X3 BT225565-300X6
Molybdenum ≥99.95% (trace metals basis), foil, Thickness 0.127 mm
Molybdenum
CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465; RTECS: QA4680000 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3

  • RTECS: QA4680000
  • Solid


Formula: Mo
Boiling Pt: 4612 °C
Melting Pt: 2617 °C
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty