Short Form High Precision Specific Gravity Hydrometers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
52112-0809 52112-0909 52112-0910 52112-1011 52112-1111 52112-1112 52112-1113 52112-1213
10169-814EA 46.27 USD InStock
10169-814 10169-816 10169-818 10169-820 10169-822 10169-824 10169-826 10169-828
Short Form High Precision Specific Gravity Hydrometers, Kimble Chase
Hydrometers
Serial number is printed on the scale and a certificate of traceability is included
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty