Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD3 (T Cell) Rabbit Monoclonal Antibody

Supplier: Diagnostic Biosystems
RP005 RP005-05
99991-208EA 642.4 USD InStock
99991-208 95024-936
Anti-CD3 (T Cell) Rabbit Monoclonal Antibody
Antibodies
Type:
Antigen: CD3 (T Cell)
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty