Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD51/61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 23C6]

Supplier: Biolegend
304403 304404
304403-BLEA 93.4 USD InStock
304403-BL 304404-BL
Anti-CD51/61 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 23C6]
Antibodies
FITC anti-human CD51/61 [23C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Chicken, Cynomolgus, Rabbit (Lapine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD51/61
Clonality: Monoclonal
Clone: 23C6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Chicken;Cynomolgus;Rabbit (Lapine)

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty