Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-H-2Kb/H-2Db Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 28-8-6]

Supplier: Biolegend
114605 114606
114605-BLEA 105.8 USD InStock
114605-BL 114606-BL
Anti-H-2Kb/H-2Db Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 28-8-6]
Antibodies
FITC anti-mouse H-2Kb/H-2Db [28-8-6]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2Dd MHC class I alloantigen. Does not react with other haplotypes (e.g., f, k, p, q, r, s); Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb/H-2Db
Clonality: Monoclonal
Clone: 28-8-6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse (C3H) IgG2a, κ
Reactivity: Mouse b haplotype;Cross-reacts with H-2Dd MHC class I alloantigen. Does not react with other haplotypes (e.g.;f;k;p;q;r;s)

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty