Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RTK2118]

Supplier: Biolegend
400805 400806
400805-BLEA 97.66 USD InStock
400805-BL 400806-BL
Anti-IgM, k Isotype Ctrl Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RTK2118]
Antibodies
FITC Rat IgM, κ Isotype Ctrl [RTK2118]; Isotype: Rat IgM, κ; Reactivity: Rat; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgM, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2118
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty