Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Acetylated Lysine Mouse Monoclonal Antibody [clone: 15G10]

Supplier: Biolegend
623401 623402
623401-BLEA 118.86 USD InStock
623401-BL 623402-BL
Anti-Acetylated Lysine Mouse Monoclonal Antibody [clone: 15G10]
Antibodies
Purified anti-Acetylated Lysine [15G10]; Isotype: Mouse IgG2b, κ; Reactivity: Acetylated lysine residues; Apps: WB, IF; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy: Use a dilution range of 1-4 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Acetylated Lysine
Clonality: Monoclonal
Clone: 15G10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Acetylated lysine residues

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty