Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Supplier: Biotium
20170 20170-1
97065-528EA 260.49 USD InStock
97065-528 97065-530
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Guinea Pig

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty