Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Ki-67 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: Ki-67]

Supplier: Biolegend
350505 350506
350505-BLEA 246.2 USD InStock
350505-BL 10019-446
Anti-Ki-67 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: Ki-67]
Antibodies
Brilliant Violet 421™ anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC, IF; Size: 100 tests

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: Ki-67
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty