Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminothiophenol ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

A0623-25G A0623-5G
TCA0623-25GEA 108 USD InStock
TCA0623-25G TCA0623-5G
4-Aminothiophenol ≥98.0% (by GC)
4-Aminothiophenol
CAS Number: 1193-02-8
MDL Number: MFCD00007880
Molecular Formula: C6H7NS
Molecular Weight: 125.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 142
Flash Point (°C): 113
Freezing point (°C): 37
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 4-Aminothiophenol, 4-Mercaptoaniline


Formula: C₆H₇NS
Boiling Pt: 142 °C
Flash Pt: 113 °C
MDL Number: MFCD00007880
CAS Number: 1193-02-8
UN: 3263
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a673105207e0bd05f516ff1f37a1f86e191fc6&vid=18a673105207e0bd05f516ff1f37a1f86e191fc6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty