Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsilyl acetate (acetoxytrimethylsilane) ≥98.0%

Supplier: Decon Laboratories Ltd

A1567-250ML A1567-25ML
TCA1567-250MLEA 1430.1 USD InStock
TCA1567-250ML TCA1567-25ML
Trimethylsilyl acetate (acetoxytrimethylsilane) ≥98.0%
Trimethylsilyl acetate (acetoxytrimethylsilane)
CAS Number: 2754-27-0
MDL Number: MFCD00008695
Molecular Formula: C5H12O2Si
Molecular Weight: 132.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Melting point (°C): -32
Flash Point (°C): 4
Specific Gravity (20/20): 0.88

Other Product Names / Synonyms: Acetic Acid Trimethylsilyl Ester


Formula: C₅H₁₂O₂Si
MW: 132.24 g/mol
Boiling Pt: 108 °C
Melting Pt: –32 °C
Density: 0.88 (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00008695
CAS Number: 2754-27-0
UN: 3272
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a9de1b89c9f5ac28973daf6ee665d658b5daa25&vid=9a9de1b89c9f5ac28973daf6ee665d658b5daa25&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty