A1591-25ML
A1591-5ML
TCA1591-25MLEA
121.06
USD
InStock
TCA1591-25ML
TCA1591-5ML
4-Aminopiperidine ≥96.0% (by GC, titration analysis)
4-Aminopiperidine
CAS Number: 13035-19-3
MDL Number: MFCD02179399
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Boiling point (°C): 36
Flash Point (°C): 48
Freezing point (°C): 25
Other Product Names / Synonyms: 4-Piperidinamine