Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethoxymethyl acetate ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd

A1608-25G A1608-5G
TCA1608-25GEA 246 USD InStock
TCA1608-25G TCA1608-5G
Diethoxymethyl acetate ≥97.0% (by GC)
Diethoxymethyl acetate
CAS Number: 14036-06-7
MDL Number: MFCD00009229
Molecular Formula: C7H14O4
Molecular Weight: 162.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 27
Specific Gravity (20/20): 1.00

Other Product Names / Synonyms: Acetic Acid Diethoxymethyl Ester


Formula: C₇H₁₄O₄
Density: 1.00 (20 °C)
Flash Pt: 27 °C
MDL Number: MFCD00009229
CAS Number: 14036-06-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8893f6385b2f27c6ad4d3f667789b109dc08d683&vid=8893f6385b2f27c6ad4d3f667789b109dc08d683&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty