Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Amyl ethyl ether ≥98.0%

Supplier: Decon Laboratories Ltd

A1673-25G A1673-5G
TCA1673-25GEA 282.5 USD InStock
TCA1673-25G TCA1673-5G
tert-Amyl ethyl ether ≥98.0%
tert-Amyl ethyl ether
CAS Number: 919-94-8
MDL Number: MFCD01727266
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Specific Gravity (20/20): 0.77

Other Product Names / Synonyms: 1,1-Dimethylpropyl Ethyl Ether, Ethyl tert-Pentyl Ether


Formula: C₇H₁₆O
Boiling Pt: 102 °C
Density: 0.77 (20 °C)
MDL Number: MFCD01727266
CAS Number: 919-94-8
UN: 3271
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f29619280dc0238ac70ff64b04f6447fdc663fa&vid=7f29619280dc0238ac70ff64b04f6447fdc663fa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty