Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-o-toluidine ≥98.0%

Supplier: Decon Laboratories Ltd

A1959-25G A1959-5G
TCA1959-25GEA 193.5 USD InStock
TCA1959-25G TCA1959-5G
6-Chloro-o-toluidine ≥98.0%
6-Chloro-o-toluidine
CAS Number: 87-63-8
MDL Number: MFCD00007679
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 215
Melting point (°C): 12
Flash Point (°C): 105
Specific Gravity (20/20): 1.17

Other Product Names / Synonyms: 2-Amino-3-chlorotoluene, 6-Chloro-o-toluidine


Formula: C₇H₈ClN
Boiling Pt: 215 °C
Melting Pt: 12 °C
Density: 1.17 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00007679
CAS Number: 87-63-8
UN: 3429
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4268e6cbaa96f2d988ed9affe55fbc5f2b00160&vid=b4268e6cbaa96f2d988ed9affe55fbc5f2b00160&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty