Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl acetoacetate ≥95.0%

Supplier: Decon Laboratories Ltd

A1981-25G A1981-500G
TCA1981-25GEA 39 USD InStock
TCA1981-25G TCA1981-500G
Allyl acetoacetate ≥95.0%
Allyl acetoacetate
CAS Number: 1118-84-9
MDL Number: MFCD00009811
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Melting point (°C): -85
Flash Point (°C): 76
Specific Gravity (20/20): 1.04

Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Materials Chemistry, Monomers
Other Product Names / Synonyms: Acetoacetic Acid Allyl Ester


Formula: C₇H₁₀O₃
Boiling Pt: 195 °C
Melting Pt: -85 °C
Density: 1.04 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00009811
CAS Number: 1118-84-9
UN: 2810
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93da40a8f76995e39d0c0dcf15bb6e9c0bce61e9&vid=93da40a8f76995e39d0c0dcf15bb6e9c0bce61e9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty