Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-N-methylbenzylamine ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

A2158-25G A2158-5G
TCA2158-25GEA 30 USD InStock
TCA2158-25G TCA2158-5G
3-Amino-N-methylbenzylamine ≥97.0% (by GC, titration analysis)
3-Amino-N-methylbenzylamine
CAS Number: 18759-96-1
MDL Number: MFCD00211072
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Freezing point (°C): 34

Other Product Names / Synonyms: 3-(Methylaminomethyl)aniline


Formula: C₈H₁₂N₂
MDL Number: MFCD00211072
CAS Number: 18759-96-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30820f0137051c96587c1f9e7e64a6128e159ca6&vid=30820f0137051c96587c1f9e7e64a6128e159ca6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty