A2401-1G
A2401-5G
TCA2401-1GEA
57.5
USD
InStock
TCA2401-1G
TCA2401-5G
Amlexanox ≥98.0% (by HPLC, titration analysis)
Amlexanox
CAS Number: 68302-57-8
Molecular Formula: C16H14N2O4
Molecular Weight: 298.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 300
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 2-Amino-7-isopropyl-5-oxochromeno[2,3-b]pyridine-3-carboxylic Acid