Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(3-bromophenyl)propionic acid ≥98.0%

Supplier: Decon Laboratories Ltd

A2535-1G A2535-5G
TCA2535-1GEA 57.5 USD InStock
TCA2535-1G TCA2535-5G
3-Amino-3-(3-bromophenyl)propionic acid ≥98.0%
3-Amino-3-(3-bromophenyl)propionic acid
CAS Number: 117391-50-1
MDL Number: MFCD00995843
Molecular Formula: C9H10BrNO2
Molecular Weight: 244.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226


Formula: C₉H₁₀BrNO₂
MW: 244.0852 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00995843
CAS Number: 117391-50-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64de6b4c1b592089be55cf5d646f6d86985f1858&vid=64de6b4c1b592089be55cf5d646f6d86985f1858&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty