Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis), high purity

Supplier: Decon Laboratories Ltd

A5721-1G A5721-5G
TCA5721-1GEA 90 USD InStock
TCA5721-1G TCA5721-5G
Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis), high purity
Perfluoroheptanoic acid
[Ion-Pair Reagent for LC-MS]
CAS Number: 375-85-9
MDL Number: MFCD00039604
Molecular Formula: C7HF13O2
Molecular Weight: 364.06
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 177
Flash Point (°C): 110
Freezing point (°C): 34

Product Categories: Analytical Science, LC/MS Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: Perfluoroheptanoic Acid High Grade, IPC-PFFA-7 HG


Formula: C₇HF₁₃O₂
Boiling Pt: 177 °C
Flash Pt: 110 °C
MDL Number: MFCD00039604
CAS Number: 375-85-9
UN: 3261
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b639be773c096ebc748bf581407c2d59f1c1bcea&vid=b639be773c096ebc748bf581407c2d59f1c1bcea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty