Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-1,1'-Bi-2-naphthol ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

B0461-25G B0461-5G
TCB0461-25GEA 84 USD InStock
TCB0461-25G TCB0461-5G
(±)-1,1'-Bi-2-naphthol ≥99.0% (by GC)
(±)-1,1'-Bi-2-naphthol
CAS Number: 602-09-5
MDL Number: MFCD00004068
Molecular Formula: C20H14O2
Molecular Weight: 286.33
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 220

Other Product Names / Synonyms: 2,2'-Dihydroxy-1,1'-binaphthyl


Formula: C₂₀H₁₄O₂
Melting Pt: 220 °C
MDL Number: MFCD00004068
CAS Number: 602-09-5
UN: 2811
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0a229d46ff73741124a2deaf8abb9356b68485e&vid=f0a229d46ff73741124a2deaf8abb9356b68485e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty