Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-α,α,α-trifluorotoluene ≥98.0%

Supplier: Decon Laboratories Ltd

B0663-250G B0663-25G
TCB0663-250GEA 142 USD InStock
TCB0663-250G TCB0663-25G
2-Bromo-α,α,α-trifluorotoluene ≥98.0%
2-Bromo-α,α,α-trifluorotoluene
CAS Number: 392-83-6
MDL Number: MFCD00000373
Molecular Formula: C7H4BrF3
Molecular Weight: 225.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 51
Specific Gravity (20/20): 1.67

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-alpha,alpha,alpha-trifluorotoluene


Formula: C₇H₄BrF₃
Boiling Pt: 171 °C
Density: 1.67 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00000373
CAS Number: 392-83-6
UN: 1993
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6a61d364aa4d03a9585991c6490e2fa70cadbf0&vid=d6a61d364aa4d03a9585991c6490e2fa70cadbf0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty