Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

B1003-25G B1003-5G
TCB1003-25GEA 97 USD InStock
TCB1003-25G TCB1003-5G
4-Butoxyaniline ≥98.0% (by GC, titration analysis)
4-Butoxyaniline
CAS Number: 4344-55-2
MDL Number: MFCD00007866
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 110
Specific Gravity (20/20): 1.01


Formula: C₁₀H₁₅NO
Boiling Pt: 157 °C
Density: 1.01 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00007866
CAS Number: 4344-55-2
UN: 2810
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d17aeaab7dee9c0946b0160a656dea4b7b29705a&vid=d17aeaab7dee9c0946b0160a656dea4b7b29705a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty