Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(trimethylsilyl) sulfate ≥98.0%

Supplier: Decon Laboratories Ltd

B1245-25G B1245-5G
TCB1245-25GEA 235 USD InStock
TCB1245-25G TCB1245-5G
Bis(trimethylsilyl) sulfate ≥98.0%
Bis(trimethylsilyl) sulphate
CAS Number: 18306-29-1 MDL Number: MFCD00008268 Molecular Formula: C6H18O4SSi2 Molecular Weight: 242.44 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 90 Melting point (°C): 46 Flash Point (°C): -2


Formula: C₆H₁₈O₄SSi₂
MW: 242.4407 g/mol
Boiling Pt: 90 °C
Melting Pt: 46 °C
Flash Pt: –2 °C
MDL Number: MFCD00008268
CAS Number: 18306-29-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca2b071ad5d2f471b4d363203c7f1db968ec1a0c&vid=ca2b071ad5d2f471b4d363203c7f1db968ec1a0c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty