Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromothiophenol ≥97.0%

Supplier: Decon Laboratories Ltd

B1385-25G B1385-5G
TCB1385-25GEA 99 USD InStock
TCB1385-25G TCB1385-5G
4-Bromothiophenol ≥97.0%
4-Bromothiophenol
CAS Number: 106-53-6
MDL Number: MFCD00004845
Molecular Formula: C6H5BrS
Molecular Weight: 189.07
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 231
Melting point (°C): 74

Other Product Names / Synonyms: 4-Bromothiophenol


Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 239
Melting Pt: 74 °C
MDL Number: MFCD00004845
CAS Number: 106-53-6
UN: 2923
ADR: 8 III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e41df16c14b9503d79b71f07eaf4893ad25150&vid=52e41df16c14b9503d79b71f07eaf4893ad25150&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty