Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Benzylmaleimide ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

B1805-25G B1805-5G
TCB1805-25GEA 106 USD InStock
TCB1805-25G TCB1805-5G
N-Benzylmaleimide ≥98.0% (by HPLC)
N-Benzylmaleimide
CAS Number: 1631-26-1
MDL Number: MFCD00014540
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 70


Formula: C₁₁H₉NO₂
Boiling Pt: 177 °C
Melting Pt: 70 °C
MDL Number: MFCD00014540
CAS Number: 1631-26-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb489c1c8e7ec8eec4be824a3bb97ce396a17c55&vid=cb489c1c8e7ec8eec4be824a3bb97ce396a17c55&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty