Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiophenol ≥98.0%

Supplier: Decon Laboratories Ltd

B2347-25G B2347-5G
TCB2347-25GEA 158.67 USD InStock
TCB2347-25G TCB2347-5G
2-Bromothiophenol ≥98.0%
2-Bromothiophenol
CAS Number: 6320-02-1
MDL Number: MFCD00004827
Molecular Formula: C6H5BrS
Molecular Weight: 189.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 110
Specific Gravity (20/20): 1.61
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 2-Bromothiophenol


Formula: C₆H₅BrS
Boiling Pt: 93 °C
Density: 1.61 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004827
CAS Number: 6320-02-1
UN: 2810
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f30371d5b047f601d794a9fa05935ddc356121&vid=e2f30371d5b047f601d794a9fa05935ddc356121&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty