Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobenzonitrile ≥98.0%

Supplier: Decon Laboratories Ltd

B2745-25G B2745-5G
TCB2745-25GEA 181 USD InStock
TCB2745-25G TCB2745-5G
4-Bromo-2-fluorobenzonitrile ≥98.0%
4-Bromo-2-fluorobenzonitrile
CAS Number: 105942-08-3
Molecular Formula: C7H3BrFN
Molecular Weight: 200.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 239
Melting point (°C): 71


Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 239 °C
Melting Pt: 71 °C
MDL Number: MFCD00143264
CAS Number: 105942-08-3
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty