Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[Bis(4-fluorophenyl)methyl]piperazine ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd

B2764-25G B2764-5G
TCB2764-25GEA 200.9 USD InStock
TCB2764-25G TCB2764-5G
1-[Bis(4-fluorophenyl)methyl]piperazine ≥97.0% (by GC)
1-[Bis(4-fluorophenyl)methyl]piperazine
CAS Number: 27469-60-9
Molecular Formula: C17H18F2N2
Molecular Weight: 288.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 93

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(4,4'-Difluorobenzhydryl)piperazine, 1-(4,4'-Difluorodiphenylmethyl)piperazine


Formula: C₁₇H₁₈F₂N₂
Boiling Pt: 130 °C
Melting Pt: 93 °C
MDL Number: MFCD00038660
CAS Number: 27469-60-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0ca71626c1b03206cf4dcb84ed8d6f99f5b10cf&vid=c0ca71626c1b03206cf4dcb84ed8d6f99f5b10cf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty